Anti-SPPL2A Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPPL2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-559
- 77833-558
- Antigen Symbol:
- SPPL2A
- SPPL2A
- Antigen Name:
- Signal peptide peptidase-like 2A protein (26-172AA)
- Signal peptide peptidase-like 2A protein (26-172AA)
- Antigen Synonyms:
- SPPL2A|IMP3|PSL2|PSEC0147|Signal peptide peptidase-like 2A|SPP-like 2A|EC 3.4.23.-|Intramembrane protease 3|IMP-3|Presenilin-like protein 2
- SPPL2A|IMP3|PSL2|PSEC0147|Signal peptide peptidase-like 2A|SPP-like 2A|EC 3.4.23.-|Intramembrane protease 3|IMP-3|Presenilin-like protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TCT8
- Q8TCT8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice