Anti-SEC11A Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SEC11A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77798-959
- 77798-960
- Antigen Symbol:
- SEC11A
- SEC11A
- Antigen Name:
- Signal peptidase complex catalytic subunit SEC11A protein (37-179AA)
- Signal peptidase complex catalytic subunit SEC11A protein (37-179AA)
- Antigen Synonyms:
- 1810012E07Rik|Endopeptidase SP18|Microsomal signal peptidase 18 kDa subunit|SC11A|SC11A_HUMAN|SEC11 homolog A (S. cerevisiae)|SEC11 homolog A|SEC11-like protein 1|SEC11A|SEC11L1|sid2895|Signal peptidase complex (18kD)|Signal peptidase complex catalytic subunit SEC11A|SPase 18 kDa subunit|SPC18|SPCS4A
- 1810012E07Rik|Endopeptidase SP18|Microsomal signal peptidase 18 kDa subunit|SC11A|SC11A_HUMAN|SEC11 homolog A (S. cerevisiae)|SEC11 homolog A|SEC11-like protein 1|SEC11A|SEC11L1|sid2895|Signal peptidase complex (18kD)|Signal peptidase complex catalytic subunit SEC11A|SPase 18 kDa subunit|SPC18|SPCS4A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P67812
- P67812
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice