Anti-RGS20 Rabbit Recombinant Polyclonal Antibody
Catalog # 77797-213
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Regulator of G-protein signaling 20 protein (1-241AA)
- Antigen Symbol:RGS20
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O76081
- Antigen Synonyms:G(z)GAP|Gz selective GTPase activating protein|Gz-GAP|Gz-selective GTPase-activating protein|HGNC:14600|Regulator of G protein signaling 20|Regulator of G-protein signaling 20|Regulator of G-protein signaling Z1|Regulator of Gz selective protein signaling 1|Regulator of Gz-selective protein signaling 1|RGS20|RGS20_HUMAN|RGSZ1|ZGAP1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RGS20
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse