Anti-HSD3B7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HSD3B7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-493
- 77835-494
- Antigen Symbol:
- HSD3B73 beta-hydroxysteroid dehydrogenase type 7;3 beta-hydroxysteroid dehydrogenase type VII;3-beta-HSD VII;3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase;C(27) 3-beta-HSD;EC 1.1.1.-;Cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase;EC 1.1.1.181
- HSD3B73 beta-hydroxysteroid dehydrogenase type 7;3 beta-hydroxysteroid dehydrogenase type VII;3-beta-HSD VII;3-beta-hydroxy-Delta(5)-C27 steroid oxidoreductase;C(27) 3-beta-HSD;EC 1.1.1.-;Cholest-5-ene-3-beta,7-alpha-diol 3-beta-dehydrogenase;EC 1.1.1.181
- Antigen Name:
- 3 beta-hydroxysteroid dehydrogenase type 7 protein (131-197AA)
- 3 beta-hydroxysteroid dehydrogenase type 7 protein (131-197AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H2F3
- Q9H2F3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice