Anti-USP48 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP48
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-695
- 77832-694
- Antigen Symbol:
- USP48
- USP48
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 48 protein (358-508AA)
- Ubiquitin carboxyl-terminal hydrolase 48 protein (358-508AA)
- Antigen Synonyms:
- AAH67261|Deubiquitinating enzyme 48|DKFZp762M1713|EC 3.1.2.15|Kiaa4202|MGC132556|MGC14879|RAP1GA1|Synaptic ubiquitin specific protease|synUSP|Ubiquitin carboxyl terminal hydrolase 48|Ubiquitin carboxyl-terminal hydrolase 48|Ubiquitin specific peptidase 48|Ubiquitin specific processing protease 48|Ubiquitin specific protease 31|Ubiquitin specific protease 48|Ubiquitin thioesterase 48|Ubiquitin thiolesterase 48|Ubiquitin-specific-processing protease 48|UBP48_HUMAN|USP31|USP48
- AAH67261|Deubiquitinating enzyme 48|DKFZp762M1713|EC 3.1.2.15|Kiaa4202|MGC132556|MGC14879|RAP1GA1|Synaptic ubiquitin specific protease|synUSP|Ubiquitin carboxyl terminal hydrolase 48|Ubiquitin carboxyl-terminal hydrolase 48|Ubiquitin specific peptidase 48|Ubiquitin specific processing protease 48|Ubiquitin specific protease 31|Ubiquitin specific protease 48|Ubiquitin thioesterase 48|Ubiquitin thiolesterase 48|Ubiquitin-specific-processing protease 48|UBP48_HUMAN|USP31|USP48
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UV5
- Q86UV5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice