Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP48 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-USP48 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP48 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77832-697
  • 77832-696
  • Antigen Symbol:
  • USP48
  • USP48
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 48 protein (358-508AA)
  • Ubiquitin carboxyl-terminal hydrolase 48 protein (358-508AA)
  • Antigen Synonyms:
  • AAH67261|Deubiquitinating enzyme 48|DKFZp762M1713|EC 3.1.2.15|Kiaa4202|MGC132556|MGC14879|RAP1GA1|Synaptic ubiquitin specific protease|synUSP|Ubiquitin carboxyl terminal hydrolase 48|Ubiquitin carboxyl-terminal hydrolase 48|Ubiquitin specific peptidase 48|Ubiquitin specific processing protease 48|Ubiquitin specific protease 31|Ubiquitin specific protease 48|Ubiquitin thioesterase 48|Ubiquitin thiolesterase 48|Ubiquitin-specific-processing protease 48|UBP48_HUMAN|USP31|USP48
  • AAH67261|Deubiquitinating enzyme 48|DKFZp762M1713|EC 3.1.2.15|Kiaa4202|MGC132556|MGC14879|RAP1GA1|Synaptic ubiquitin specific protease|synUSP|Ubiquitin carboxyl terminal hydrolase 48|Ubiquitin carboxyl-terminal hydrolase 48|Ubiquitin specific peptidase 48|Ubiquitin specific processing protease 48|Ubiquitin specific protease 31|Ubiquitin specific protease 48|Ubiquitin thioesterase 48|Ubiquitin thiolesterase 48|Ubiquitin-specific-processing protease 48|UBP48_HUMAN|USP31|USP48
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q86UV5
  • Q86UV5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications