Anti-USP4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: USP4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-371
- 77822-370
- Antigen Symbol:
- USP4
- USP4
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase 4 protein (632-810AA)
- Ubiquitin carboxyl-terminal hydrolase 4 protein (632-810AA)
- Antigen Synonyms:
- Deubiquitinating enzyme 4|MGC149848|MGC149849|Proto oncogene|Protooncogene|Ubiquitin carboxyl terminal hydrolase 4|Ubiquitin carboxyl-terminal hydrolase 4|Ubiquitin specific peptidase 4 (proto oncogene)|Ubiquitin specific peptidase 4 (protooncogene)|Ubiquitin specific peptidase 4|Ubiquitin specific processing protease 4|Ubiquitin specific protease 4 (proto oncogene)|Ubiquitin specific protease 4 (protooncogene)|Ubiquitin specific protease 4|Ubiquitin specific protease proto oncogene|Ubiquitin specific protease protooncogene|Ubiquitin thioesterase 4|Ubiquitin thiolesterase 4|Ubiquitin-specific-processing protease 4|Ubiquitous nuclear protein homolog|UBP4_HUMAN|UNP|UNPH|USP 4|USP4
- Deubiquitinating enzyme 4|MGC149848|MGC149849|Proto oncogene|Protooncogene|Ubiquitin carboxyl terminal hydrolase 4|Ubiquitin carboxyl-terminal hydrolase 4|Ubiquitin specific peptidase 4 (proto oncogene)|Ubiquitin specific peptidase 4 (protooncogene)|Ubiquitin specific peptidase 4|Ubiquitin specific processing protease 4|Ubiquitin specific protease 4 (proto oncogene)|Ubiquitin specific protease 4 (protooncogene)|Ubiquitin specific protease 4|Ubiquitin specific protease proto oncogene|Ubiquitin specific protease protooncogene|Ubiquitin thioesterase 4|Ubiquitin thiolesterase 4|Ubiquitin-specific-processing protease 4|Ubiquitous nuclear protein homolog|UBP4_HUMAN|UNP|UNPH|USP 4|USP4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13107
- Q13107
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together