Anti-MACROD1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MACROD1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-994
- 77834-995
- Antigen Symbol:
- MACROD1
- MACROD1
- Antigen Name:
- O-acetyl-ADP-ribose deacetylase MACROD1 protein (83-325AA)
- O-acetyl-ADP-ribose deacetylase MACROD1 protein (83-325AA)
- Antigen Synonyms:
- MACROD1|LRP16|ADP-ribose glycohydrolase MACROD1|MACRO domain-containing protein 1|O-acetyl-ADP-ribose deacetylase MACROD1|EC 3.1.1.106|Protein LRP16|[Protein ADP-ribosylaspartate] hydrolase MACROD1|EC 3.2.2.-|[Protein ADP-ribosylglutamate] hydrolase MACROD1
- MACROD1|LRP16|ADP-ribose glycohydrolase MACROD1|MACRO domain-containing protein 1|O-acetyl-ADP-ribose deacetylase MACROD1|EC 3.1.1.106|Protein LRP16|[Protein ADP-ribosylaspartate] hydrolase MACROD1|EC 3.2.2.-|[Protein ADP-ribosylglutamate] hydrolase MACROD1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BQ69
- Q9BQ69
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice