Anti-RSPH3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RSPH3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-938
- 77831-939
- Antigen Symbol:
- RSPH3
- RSPH3
- Antigen Name:
- Radial spoke head protein 3 homolog protein (1-275AA)
- Radial spoke head protein 3 homolog protein (1-275AA)
- Antigen Synonyms:
- RSPH3|RSHL2|RSP3|Radial spoke head protein 3 homolog|A-kinase anchor protein RSPH3|Radial spoke head-like protein 2
- RSPH3|RSHL2|RSP3|Radial spoke head protein 3 homolog|A-kinase anchor protein RSPH3|Radial spoke head-like protein 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UC2
- Q86UC2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice