Anti-CYLD Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CYLD
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-432
- 77836-433
- Antigen Symbol:
- CYLD
- CYLD
- Antigen Name:
- Ubiquitin carboxyl-terminal hydrolase CYLD protein (269-435AA)
- Ubiquitin carboxyl-terminal hydrolase CYLD protein (269-435AA)
- Antigen Synonyms:
- BRSS|CDMT|CYLD gene|CYLD_HUMAN|CYLD1|CYLDI|cylindromatosis (turban tumor syndrome)|cylindromatosis 1|Deubiquitinating enzyme EAC|HSPC057|KIAA0849|MFT|MFT1|Probable ubiquitin carboxyl terminal hydrolase SBS|TEM|turban tumor syndrome|Ubiquitin carboxyl-terminal hydrolase ubiquitin specific peptidase like 2|ubiquitin thioesterase Ubiquitin thiolesterase Ubiquitin-specific processing protease Ubiquitin-specific-processing protease USPL2
- BRSS|CDMT|CYLD gene|CYLD_HUMAN|CYLD1|CYLDI|cylindromatosis (turban tumor syndrome)|cylindromatosis 1|Deubiquitinating enzyme EAC|HSPC057|KIAA0849|MFT|MFT1|Probable ubiquitin carboxyl terminal hydrolase SBS|TEM|turban tumor syndrome|Ubiquitin carboxyl-terminal hydrolase ubiquitin specific peptidase like 2|ubiquitin thioesterase Ubiquitin thiolesterase Ubiquitin-specific processing protease Ubiquitin-specific-processing protease USPL2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NQC7
- Q9NQC7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together