Anti-POLDIP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POLDIP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-131
- 77838-130
- Antigen Symbol:
- POLDIP3
- POLDIP3
- Antigen Name:
- Polymerase delta-interacting protein 3 protein (100-221AA)
- Polymerase delta-interacting protein 3 protein (100-221AA)
- Antigen Synonyms:
- 46 kDa DNA polymerase delta interaction protein|DNA polymerase delta interacting protein 3|KIAA1649|P46|PDIP3_HUMAN|PDIP46|POLDIP3|Polymerase (DNA-directed), delta interacting protein 3|Polymerase delta interacting protein 46|Polymerase delta-interacting protein 3|RNA-binding protein P46|S6K1 Aly/REF-like target|SKAR
- 46 kDa DNA polymerase delta interaction protein|DNA polymerase delta interacting protein 3|KIAA1649|P46|PDIP3_HUMAN|PDIP46|POLDIP3|Polymerase (DNA-directed), delta interacting protein 3|Polymerase delta interacting protein 46|Polymerase delta-interacting protein 3|RNA-binding protein P46|S6K1 Aly/REF-like target|SKAR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BY77
- Q9BY77
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice