Anti-TTC27 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TTC27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-869
- 77830-870
- Antigen Symbol:
- TTC27
- TTC27
- Antigen Name:
- Tetratricopeptide repeat protein 27 protein (590-817AA)
- Tetratricopeptide repeat protein 27 protein (590-817AA)
- Antigen Synonyms:
- FLJ20272|Tetratricopeptide repeat domain 27|Tetratricopeptide repeat protein 27|TPR repeat protein 27|TTC27|TTC27_HUMAN
- FLJ20272|Tetratricopeptide repeat domain 27|Tetratricopeptide repeat protein 27|TPR repeat protein 27|TTC27|TTC27_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P3X3
- Q6P3X3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice