Anti-MED27 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: MED27
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77831-403
- 77831-402
- Antigen Symbol:
- MED27
- MED27
- Antigen Name:
- Mediator of RNA polymerase II transcription subunit 27 protein (39-108AA)
- Mediator of RNA polymerase II transcription subunit 27 protein (39-108AA)
- Antigen Synonyms:
- Cofactor required for Sp1 transcriptional activation subunit 8 (34kD)|Cofactor required for Sp1 transcriptional activation subunit 8 34kDa|Cofactor required for Sp1 transcriptional activation subunit 8|CRAP 34|CRAP34|CRSP 34|CRSP 8|CRSP complex subunit 8|CRSP34|CRSP8|MED 27|Med27|MED27_HUMAN|Mediator complex subunit 27|Mediator of RNA polymerase II transcription subunit 27|MGC11274|P37 TRAP/SMCC/PC2 subunit|Transcriptional coactivator CRSP34|TRAP 37|TRAP37
- Cofactor required for Sp1 transcriptional activation subunit 8 (34kD)|Cofactor required for Sp1 transcriptional activation subunit 8 34kDa|Cofactor required for Sp1 transcriptional activation subunit 8|CRAP 34|CRAP34|CRSP 34|CRSP 8|CRSP complex subunit 8|CRSP34|CRSP8|MED 27|Med27|MED27_HUMAN|Mediator complex subunit 27|Mediator of RNA polymerase II transcription subunit 27|MGC11274|P37 TRAP/SMCC/PC2 subunit|Transcriptional coactivator CRSP34|TRAP 37|TRAP37
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6P2C8
- Q6P2C8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together