Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MSRA Rabbit Recombinant Polyclonal Antibody
Anti-MSRA Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MSRA Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77841-621
  • 77841-622
  • Antigen Symbol:
  • MSRA
  • MSRA
  • Antigen Name:
  • Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
  • Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
  • Antigen Synonyms:
  • Cytosolic methionine S sulfoxide reductase|Methionine sulphoxide reductase A|Mitochondrial peptide methionine sulfoxide reductase|MSR A|msrA|MSRA_HUMAN|peptide met (O) reductase|Peptide Met(O) reductase|Peptide methionine (S) S oxide reductase|Peptide-methionine (S)-S-oxide reductase|PMSR|Protein methionine S oxide reductase|Protein-methionine-S-oxide reductase
  • Cytosolic methionine S sulfoxide reductase|Methionine sulphoxide reductase A|Mitochondrial peptide methionine sulfoxide reductase|MSR A|msrA|MSRA_HUMAN|peptide met (O) reductase|Peptide Met(O) reductase|Peptide methionine (S) S oxide reductase|Peptide-methionine (S)-S-oxide reductase|PMSR|Protein methionine S oxide reductase|Protein-methionine-S-oxide reductase
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UJ68
  • Q9UJ68
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications