Anti-CHST6 Rabbit Polyclonal Antibody
Catalog # 77806-680
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of CHST6
- Antigen Symbol:CHST6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µl
- Form:Liquid
- Gene ID:Q9GZX3
- Antigen Synonyms:C GlcNAc6ST|C-GlcNAc6ST|Carbohydate sulfotransferase 6|Carbohydrate (N acetylglucosamine 6 O) sulfotransferase 6|Carbohydrate sulfotransferase 6|CHST6|CHST6_HUMAN|Corneal GlcNAc6-sulfotransferase|Corneal N acetylglucosamine 6 sulfotransferase|Corneal N-acetylglucosamine-6-O-sulfotransferase|Galactose N acetylglucosamine N acetylglucosamine 6 O sulfotransferase 4 beta|Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta|GlcNAc6ST 5|GlcNAc6ST-5|Gn6st-5|GST4 beta|GST4-beta|hCGn6ST|N-acetylglucosamine 6-O-sulfotransferase 5
- Storage Buffer:PBS, 150 mM NaCl, 0.02% Sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:100 to 1:500
Type:
Antigen: CHST6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human