Anti-Tpsb2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: Tpsb2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77803-410
- 77811-410
- Antigen Symbol:
- Tpsb2
- Tpsb2
- Antigen Name:
- Recombinant Mouse Tryptase beta-2 protein (32-276AA)
- Recombinant Mouse Tryptase beta-2 protein (32-276AA)
- Antigen Synonyms:
- Tpsb2|Mcpt6|Tryptase beta-2|Tryptase-2|EC 3.4.21.59|Mast cell protease 6|mMCP-6
- Tpsb2|Mcpt6|Tryptase beta-2|Tryptase-2|EC 3.4.21.59|Mast cell protease 6|mMCP-6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21845
- P21845
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice