Anti-LRRN1 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: LRRN1
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-122
- 77830-123
- Antigen Symbol:
- LRRN1
- LRRN1
- Antigen Name:
- Leucine-rich repeat neuronal protein 1 protein (26-631AA)
- Leucine-rich repeat neuronal protein 1 protein (26-631AA)
- Antigen Synonyms:
- FIGLER3|KIAA1497|Leucine-rich repeat neuronal protein 1|Lrrn1|LRRN1_HUMAN|Nbla10449|Neuronal leucine-rich repeat protein 1|NLRR-1
- FIGLER3|KIAA1497|Leucine-rich repeat neuronal protein 1|Lrrn1|LRRN1_HUMAN|Nbla10449|Neuronal leucine-rich repeat protein 1|NLRR-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UXK5
- Q6UXK5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice