Anti-EAF2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77833-989
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:ELL-associated factor 2 protein (6-100AA)
- Antigen Symbol:BM 040;BM040;EAF 2;EAF2;EAF2_HUMAN;Ehrlich S II transcriptional activator factor;ELL associated factor 2;ELL-associated factor 2;Festa;Testosterone regulated apoptosis inducer and tumor suppressor;Testosterone regulated apoptosis inducer and tumor suppressor protein;Testosterone-regulated apoptosis inducer and tumor suppressor protein;TRAITS;U19;Uncharacterized bone marrow protein BM 040;Uncharacterized bone marrow protein BM040
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96CJ1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:1000 to 1:2000, IF:1:50 to 1:500
Type:
Antigen: EAF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse