Anti-SLC40A1 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SLC40A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77836-355
- 77836-356
- Antigen Symbol:
- SLC40A1
- SLC40A1
- Antigen Name:
- Solute carrier family 40 member 1 protein (148-298AA)
- Solute carrier family 40 member 1 protein (148-298AA)
- Antigen Synonyms:
- Ferroportin 1|Ferroportin|Ferroportin-1|FPN1|HFE4|IREG1|iron regulated gene 1|Iron regulated transporter 1|Iron-regulated transporter 1|MST079|MSTP079|MTP1|putative ferroportin 1 variant IIIA|putative ferroportin 1 variant IIIB|S40A1_HUMAN|SLC11A3|formerly|solute carrier family 11 (proton-coupled divalent metal ion transporters)|member 3|solute carrier family 40 (iron-regulated transporter)|member 1|Solute carrier family 40 member 1
- Ferroportin 1|Ferroportin|Ferroportin-1|FPN1|HFE4|IREG1|iron regulated gene 1|Iron regulated transporter 1|Iron-regulated transporter 1|MST079|MSTP079|MTP1|putative ferroportin 1 variant IIIA|putative ferroportin 1 variant IIIB|S40A1_HUMAN|SLC11A3|formerly|solute carrier family 11 (proton-coupled divalent metal ion transporters)|member 3|solute carrier family 40 (iron-regulated transporter)|member 1|Solute carrier family 40 member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NP59
- Q9NP59
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice