Anti-XAGE1A Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: XAGE1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-577
- 77838-578
- Antigen Symbol:
- XAGE1A
- XAGE1A
- Antigen Name:
- X antigen family member 1 protein (1-81AA)
- X antigen family member 1 protein (1-81AA)
- Antigen Synonyms:
- Cancer/testis antigen 12.1|CT12.1|G antigen family D 2 protein|G antigen family D member 2|GAGD2_HUMAN|GAGED2|Protein XAGE-1|X antigen family, member 1|XAGE1B|XAGE1C|XAGE1D|XAGE1E
- Cancer/testis antigen 12.1|CT12.1|G antigen family D 2 protein|G antigen family D member 2|GAGD2_HUMAN|GAGED2|Protein XAGE-1|X antigen family, member 1|XAGE1B|XAGE1C|XAGE1D|XAGE1E
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HD64
- Q9HD64
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice