Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VARS2 Rabbit Recombinant Polyclonal Antibody
Anti-VARS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-VARS2 Rabbit Recombinant Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77829-125
  • 77829-126
  • Antigen Symbol:
  • VARS2
  • VARS2
  • Antigen Name:
  • Valine--tRNA ligase, mitochondrial protein (830-981AA)
  • Valine--tRNA ligase, mitochondrial protein (830-981AA)
  • Antigen Synonyms:
  • COXPD20|mitochondrial|SYVM_HUMAN|Valine--tRNA ligase|ValRS|Valyl tRNA synthetase 2 mitochondrial|Valyl-tRNA synthetase|Valyl-tRNA synthetase-like|Vars2|VARS2L|VARSL
  • COXPD20|mitochondrial|SYVM_HUMAN|Valine--tRNA ligase|ValRS|Valyl tRNA synthetase 2 mitochondrial|Valyl-tRNA synthetase|Valyl-tRNA synthetase-like|Vars2|VARS2L|VARSL
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q5ST30
  • Q5ST30
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications