Anti-PRKAA1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PRKAA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77823-390
- 77823-389
- Antigen Symbol:
- PRKAA1
- PRKAA1
- Antigen Name:
- 5\\\'-AMP-activated protein kinase catalytic subunit alpha-1 protein (325-543AA)
- 5\\\'-AMP-activated protein kinase catalytic subunit alpha-1 protein (325-543AA)
- Antigen Synonyms:
- 5 AMP activated protein kinase alpha 1catalytic subunit|5 AMP activated protein kinase catalytic alpha 1 chain|5' AMP activated protein kinase catalytic subunit alpha 1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AAPK1|AAPK1_HUMAN|ACACA kinase|acetyl CoA carboxylase kinase|AI194361|AI450832|AL024255|AMP -activate kinase alpha 1 subunit|AMP-activate kinase alpha 1 subunit |AMP-activated protein kinase, catalytic, alpha -1|AMPK 1|AMPK alpha 1|AMPK alpha 1 chain|AMPK|AMPK subunit alpha 1|AMPK subunit alpha-1|AMPK1|AMPKa1|AMPKalpha1|C130083N04Rik|cb116|EC 2.7.11.1|HMG CoA reductase kinase|HMGCR kinase|hormone sensitive lipase kinase|Hydroxymethylglutaryl CoA reductase kinase|im:7154392|kinase AMPK alpha1|MGC33776|MGC57364|OTTHUMP00000161795|OTTHUMP00000161796|PRKAA 1|Protein kinase AMP activated alpha 1 catalytic subunit|SNF1-like protein AMPK|SNF1A|Tau protein kinase wu:fa94c10
- 5 AMP activated protein kinase alpha 1catalytic subunit|5 AMP activated protein kinase catalytic alpha 1 chain|5' AMP activated protein kinase catalytic subunit alpha 1|5'-AMP-activated protein kinase catalytic subunit alpha-1|AAPK1|AAPK1_HUMAN|ACACA kinase|acetyl CoA carboxylase kinase|AI194361|AI450832|AL024255|AMP -activate kinase alpha 1 subunit|AMP-activate kinase alpha 1 subunit |AMP-activated protein kinase, catalytic, alpha -1|AMPK 1|AMPK alpha 1|AMPK alpha 1 chain|AMPK|AMPK subunit alpha 1|AMPK subunit alpha-1|AMPK1|AMPKa1|AMPKalpha1|C130083N04Rik|cb116|EC 2.7.11.1|HMG CoA reductase kinase|HMGCR kinase|hormone sensitive lipase kinase|Hydroxymethylglutaryl CoA reductase kinase|im:7154392|kinase AMPK alpha1|MGC33776|MGC57364|OTTHUMP00000161795|OTTHUMP00000161796|PRKAA 1|Protein kinase AMP activated alpha 1 catalytic subunit|SNF1-like protein AMPK|SNF1A|Tau protein kinase wu:fa94c10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13131
- Q13131
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together