Anti-TESC Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: TESC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77802-239
- 77802-238
- Antigen Symbol:
- TESC
- TESC
- Antigen Name:
- Calcineurin B homologous protein 3 protein (2-214AA)
- Calcineurin B homologous protein 3 protein (2-214AA)
- Antigen Synonyms:
- TESC|CHP3Calcineurin B homologous protein 3|Tescalcin|TSC
- TESC|CHP3Calcineurin B homologous protein 3|Tescalcin|TSC
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96BS2
- Q96BS2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice