Anti-WDR35 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WDR35
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-133
- 77840-134
- Antigen Symbol:
- WDR35
- WDR35
- Antigen Name:
- WD repeat-containing protein 35 protein (954-1181AA)
- WD repeat-containing protein 35 protein (954-1181AA)
- Antigen Synonyms:
- Intraflagellar transport protein 121 homolog|KIAA1336|MGC33196|Naofen|WD repeat domain 35|WD repeat-containing protein 35|WDR35|WDR35_HUMAN
- Intraflagellar transport protein 121 homolog|KIAA1336|MGC33196|Naofen|WD repeat domain 35|WD repeat-containing protein 35|WDR35|WDR35_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2L0
- Q9P2L0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together