Anti-RPS6KA1 Rabbit Polyclonal Antibody
Catalog # 77807-414
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from Rsk-1 around the non-phosphorylation site of T573.
- Antigen Symbol:RPS6KA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15418
- Antigen Synonyms:90 kDa ribosomal protein S6 kinase 1|dJ590P13.1 (ribosomal protein S6 kinase|90kD|polypeptide 1|dJ590P13.1|EC 2.7.11.1 |HU 1|HU1|KS6A1_HUMAN|MAP kinase activated protein kinase 1a|MAP kinase-activated protein kinase 1a|MAPK-activated protein kinase 1a|MAPKAP kinase 1a|MAPKAPK-1a|MAPKAPK1A|MGC79981|Mitogen-activated protein kinase-activated protein kinase 1A|OTTHUMP00000004113 |p90 RSK1|p90-RSK 1|p90rsk|p90RSK1|p90S6K|pp90RSK1|Ribosomal protein S6 kinase 90kD 1|Ribosomal protein S6 kinase 90kD polypeptide 1|Ribosomal protein S6 kinase 90kDa polypeptide 1|Ribosomal protein S6 kinase alpha 1|Ribosomal protein S6 kinase alpha-1|Ribosomal protein S6 kinase polypeptide 1|Ribosomal S6 kinase 1|RPS6K1 alpha|rps6ka|Rps6ka1|RSK 1|RSK 1 p90|RSK|RSK-1|RSK1|RSK1p90|S6K alpha 1 |S6K-alpha-1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:5000
Type:
Antigen: RPS6KA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat