Anti-TIMM50 Rabbit Recombinant Polyclonal Antibody
Catalog # 77826-517
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Mitochondrial import inner membrane translocase subunit TIM50 protein (87-353AA)
- Antigen Symbol:TIMM50
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q3ZCQ8
- Antigen Synonyms:Homolog of yeast Tim50|Import inner membrane translocase subunit TIM 50|Import inner membrane translocase subunit TIM50|Import inner membrane translocase subunit TIM50 mitochondrial|MGC102733|Mitochondrial import inner membrane translocase subunit TIM50|TIM 50|TIM 50L|TIM50|Tim50 like protein|TIM50_HUMAN|TIM50L|TIMM 50|TIMM50|translocase of inner mitochondrial membrane 50 homolog (S. cerevisiae)|Translocase of inner mitochondrial membrane 50 homolog (yeast)|Translocase of inner mitochondrial membrane 50 homolog|Translocase of inner mitochondrial membrane 50|yeast|homolog of
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TIMM50
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat