To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of p44S10
- Antigen Symbol:PSMD6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15008
- Antigen Synonyms:26S proteasome non-ATPase regulatory subunit 6|26S proteasome regulatory subunit RPN7|26S proteasome regulatory subunit S10|Breast cancer-associated protein SGA-113M|KIAA0107|p42A|P44S10|PFAAP4|Phosphonoformate immuno-associated protein 4|Proteasome (prosome|macropain) 26S subunit|non-ATPase|6|Proteasome regulatory particle subunit p44S10|PSMD6|PSMD6_HUMAN|Rpn7|S10|SGA-113M
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% Sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:40000
Type:
Antigen: PSMD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat