Anti-CERS6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: CERS6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77830-432
- 77830-433
- Antigen Symbol:
- CERS6
- CERS6
- Antigen Name:
- Ceramide synthase 6 protein (324-392AA)
- Ceramide synthase 6 protein (324-392AA)
- Antigen Synonyms:
- Ceramide synthase 6|CerS 6|CERS5|CerS6|LAG1 homolog ceramide synthase 6|LAG1 longevity assurance homolog 6|LASS 6|Lass6|LASS6_HUMAN|Longevity assurance homolog 6|MGC129949|MGC129950|T1L
- Ceramide synthase 6|CerS 6|CERS5|CerS6|LAG1 homolog ceramide synthase 6|LAG1 longevity assurance homolog 6|LASS 6|Lass6|LASS6_HUMAN|Longevity assurance homolog 6|MGC129949|MGC129950|T1L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6ZMG9
- Q6ZMG9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice