Anti-ABCB5 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ABCB5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-780
- 77825-779
- Antigen Symbol:
- ABCB5
- ABCB5
- Antigen Name:
- ATP-binding cassette sub-family B member 5 protein (547-760AA)
- ATP-binding cassette sub-family B member 5 protein (547-760AA)
- Antigen Synonyms:
- ABCB 5|ABCB5|ABCB5 P-gp|ABCB5_HUMAN|ABCB5alpha|ABCB5beta|ATP binding cassette protein|ATP binding cassette sub family B (MDR/TAP) member 5|ATP binding cassette sub family B member 5|ATP-binding cassette sub-family B member 5|EST422562|P glycoprotein ABCB5|P-glycoprotein ABCB5
- ABCB 5|ABCB5|ABCB5 P-gp|ABCB5_HUMAN|ABCB5alpha|ABCB5beta|ATP binding cassette protein|ATP binding cassette sub family B (MDR/TAP) member 5|ATP binding cassette sub family B member 5|ATP-binding cassette sub-family B member 5|EST422562|P glycoprotein ABCB5|P-glycoprotein ABCB5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q2M3G0
- Q2M3G0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice