Anti-KANSL3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: KANSL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77841-245
- 77841-246
- Antigen Symbol:
- KANSL3
- KANSL3
- Antigen Name:
- KAT8 regulatory NSL complex subunit 3 protein (1-200AA)
- KAT8 regulatory NSL complex subunit 3 protein (1-200AA)
- Antigen Synonyms:
- K1310_HUMAN| KAT8 regulatory NSL complex subunit 3| Kiaa1310| Non specific lethal 3 homolog| NSL complex protein NSL3| NSL3| Rcd1| Serum inhibited-related protein| Testis development protein PRTD| Uncharacterized protein KIAA1310
- K1310_HUMAN| KAT8 regulatory NSL complex subunit 3| Kiaa1310| Non specific lethal 3 homolog| NSL complex protein NSL3| NSL3| Rcd1| Serum inhibited-related protein| Testis development protein PRTD| Uncharacterized protein KIAA1310
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9P2N6
- Q9P2N6
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice