Anti-MAPK8IP2 Rabbit Recombinant Polyclonal Antibody
77824-299
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:C-Jun-amino-terminal kinase-interacting protein 2 protein (470-570AA)
- Antigen Symbol:MAPK8IP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q13387
- Antigen Synonyms:C jun amino terminal kinase interacting protein 2| C-jun-amino-terminal kinase-interacting protein 2| Homologous to mouse JIP 1| IB 2| IB-2| IB2| Islet brain 2| Islet-brain-2| JIP 2| JIP-2| JIP2| JIP2_HUMAN| JNK interacting protein 2| JNK MAP kinase scaffold protein 2| JNK MAP kinase scaffold protein JIP2| JNK-interacting protein 2| MAPK8IP2| Mitogen activated protein kinase 8 interacting protein 2| Mitogen-activated protein kinase 8-interacting protein 2| PRKM8 interacting protein like| PRKM8IPL
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: MAPK8IP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human