Anti-Adh7 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: Adh7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
- Catalog No:
- 77828-077
- 77828-078
- Antigen Symbol:
- Adh7
- Adh7
- Antigen Name:
- Recombinant Mouse Alcohol dehydrogenase class 4 mu/sigma chain protein (1-374AA)
- Recombinant Mouse Alcohol dehydrogenase class 4 mu/sigma chain protein (1-374AA)
- Antigen Synonyms:
- Adh-3|Adh3All-trans-retinol dehydrogenase [NAD(+)] ADH7|EC 1.1.1.105|ADH-C2|Alcohol dehydrogenase 7|Alcohol dehydrogenase class 4 mu/sigma chain|EC 1.1.1.1|Alcohol dehydrogenase class IV mu/sigma chain|Gastric alcohol dehydrogenase|Omega-hydroxydecanoate dehydrogenase ADH7|EC 1.1.1.66|Retinol dehydrogenase
- Adh-3|Adh3All-trans-retinol dehydrogenase [NAD(+)] ADH7|EC 1.1.1.105|ADH-C2|Alcohol dehydrogenase 7|Alcohol dehydrogenase class 4 mu/sigma chain|EC 1.1.1.1|Alcohol dehydrogenase class IV mu/sigma chain|Gastric alcohol dehydrogenase|Omega-hydroxydecanoate dehydrogenase ADH7|EC 1.1.1.66|Retinol dehydrogenase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q64437
- Q64437
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice