Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-USP47 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-USP47 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-USP47 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77834-185
  • 77834-184
  • Antigen Symbol:
  • USP47
  • USP47
  • Antigen Name:
  • Ubiquitin carboxyl-terminal hydrolase 47 protein (1-157AA)
  • Ubiquitin carboxyl-terminal hydrolase 47 protein (1-157AA)
  • Antigen Synonyms:
  • Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN|USP47
  • Deubiquitinating enzyme 47|TFRP|Trf TATA binding protein related factor proximal homolog|Trf TATA binding protein related factor proximal homolog Drosophila|Ubiquitin carboxyl-terminal hydrolase 47|Ubiquitin specific peptidase 47|Ubiquitin specific processing protease 47|Ubiquitin specific protease 47|Ubiquitin thioesterase 47|Ubiquitin thiolesterase 47|Ubiquitin-specific-processing protease 47|UBP47_HUMAN|USP47
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q96K76
  • Q96K76
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications