Anti-USP10 Rabbit Recombinant Polyclonal Antibody
Catalog # 77822-030
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 10 protein (453-592AA)
- Antigen Symbol:USP10
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q14694
- Antigen Synonyms:Deubiquitinating enzyme 10|KIAA0190|MGC124997|MGC2621|Ubiquitin carboxyl terminal hydrolase 10|Ubiquitin carboxyl-terminal hydrolase 10|Ubiquitin specific peptidase 10|Ubiquitin specific processing protease 10|Ubiquitin specific protease 10|Ubiquitin thioesterase 10|Ubiquitin thiolesterase 10|Ubiquitin-specific-processing protease 10|UBP10_HUMAN|UBPO|USP 10|USP10
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: USP10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat