Anti-IL36RN Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL36RN
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77836-703
- 77836-702
- Antigen Symbol:
- IL36RN
- IL36RN
- Antigen Name:
- Interleukin-36 receptor antagonist protein (1-155AA)
- Interleukin-36 receptor antagonist protein (1-155AA)
- Antigen Synonyms:
- AI413231|Family of interleukin 1 delta|FIL1|FIL1 delta|FIL1(DELTA)|FIL1D|Fil1delta|I36RA_HUMAN|IL 1 delta|IL 1 related protein 3|IL 1F5 (IL 1HY1, FIL1 delta, IL 1RP3, IL 1L1, IL 1 delta)|Il 1h3|IL 1HY1|IL 1L1|IL 1ra homolog|IL 1ra homolog, IL1F5 (Canonical product IL 1F5a)|IL 1RP3|IL-1 delta|IL-1-related protein 3|IL-1F5|IL-1HY1|IL-1L1|IL-1ra homolog 1|IL-1RP3|IL1F5|IL1HY1|IL1L1|IL1RP3|IL36RA|Interleukin 1 delta|Interleukin 1 family member 5 (delta)|Interleukin 1 family member 5|Interleukin 1 HY1|Interleukin 1 like protein 1|Interleukin 1 receptor antagonist homolog 1|Interleukin 36 receptor antagonist|Interleukin-1 delta|Interleukin-1 family member 5|Interleukin-1 HY1|Interleukin-1 receptor antagonist homolog 1|Interleukin-1-like protein 1|Interleukin-36 receptor antagonist protein|MGC29840|PSORP|RP23-176J12.6
- AI413231|Family of interleukin 1 delta|FIL1|FIL1 delta|FIL1(DELTA)|FIL1D|Fil1delta|I36RA_HUMAN|IL 1 delta|IL 1 related protein 3|IL 1F5 (IL 1HY1, FIL1 delta, IL 1RP3, IL 1L1, IL 1 delta)|Il 1h3|IL 1HY1|IL 1L1|IL 1ra homolog|IL 1ra homolog, IL1F5 (Canonical product IL 1F5a)|IL 1RP3|IL-1 delta|IL-1-related protein 3|IL-1F5|IL-1HY1|IL-1L1|IL-1ra homolog 1|IL-1RP3|IL1F5|IL1HY1|IL1L1|IL1RP3|IL36RA|Interleukin 1 delta|Interleukin 1 family member 5 (delta)|Interleukin 1 family member 5|Interleukin 1 HY1|Interleukin 1 like protein 1|Interleukin 1 receptor antagonist homolog 1|Interleukin 36 receptor antagonist|Interleukin-1 delta|Interleukin-1 family member 5|Interleukin-1 HY1|Interleukin-1 receptor antagonist homolog 1|Interleukin-1-like protein 1|Interleukin-36 receptor antagonist protein|MGC29840|PSORP|RP23-176J12.6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBH0
- Q9UBH0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice