Anti-MAT2B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:200 to 1:2000, IF: 1:50 to 1:200
Type:
Antigen: MAT2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Rat
Specifications
- Catalog No:
- 77841-168
- 77841-169
- Antigen Symbol:
- MAT2B
- MAT2B
- Antigen Name:
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Methionine adenosyltransferase 2 subunit beta protein (85-143AA)
- Antigen Synonyms:
- 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
- 1110064C04Rik|2410018D16Rik|AI182287|AU022853|Beta regulatory subunit of methionine adenosyltransferase|dTDP 4 keto 6 deoxy D glucose 4 reductase|DTDP-4-keto-6-deoxy-D-glucose 4-reductase|MAT II|MAT II beta|Mat2b|MAT2B_HUMAN|MATIIbeta|Methionine adenosyltransferase 2 beta subunit|Methionine adenosyltransferase 2 subunit beta|Methionine adenosyltransferase II beta|MGC12237|MSTP045|Nbla02999|OTTMUSP00000005600|Putative dTDP-4-keto-6-deoxy-D-glucose 4-reductase|Putative protein product of Nbla02999|RP23-382C18.2|SDR23E1|Short chain dehydrogenase/reductase family 23E member 1|TGR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZL9
- Q9NZL9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice