Anti-PRMT7 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PRMT7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-448
- 77842-449
- Antigen Symbol:
- PRMT7
- PRMT7
- Antigen Name:
- Protein arginine N-methyltransferase 7 protein (285-390AA)
- Protein arginine N-methyltransferase 7 protein (285-390AA)
- Antigen Synonyms:
- [Myelin basic protein]-arginine N-methyltransferase PRMT7|ANM7_HUMAN|FLJ10640|Histone-arginine N-methyltransferase PRMT7|KIAA1933|Myelin basic protein arginine N methyltransferase|OTTHUMP00000174863|PRMT7|Protein arginine methyltransferase 7|Protein arginine N-methyltransferase 7
- [Myelin basic protein]-arginine N-methyltransferase PRMT7|ANM7_HUMAN|FLJ10640|Histone-arginine N-methyltransferase PRMT7|KIAA1933|Myelin basic protein arginine N methyltransferase|OTTHUMP00000174863|PRMT7|Protein arginine methyltransferase 7|Protein arginine N-methyltransferase 7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVM4
- Q9NVM4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice