Anti-TSPAN12 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: TSPAN12
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77803-623
- 77811-623
- Antigen Symbol:
- TSPAN12
- TSPAN12
- Antigen Name:
- Tetraspanin-12 protein (111-224AA)
- Tetraspanin-12 protein (111-224AA)
- Antigen Synonyms:
- TSPAN12|NET2|TM4SF12|UNQ774/PRO1568Tetraspanin-12|Tspan-12|Tetraspan NET-2|Transmembrane 4 superfamily member 12
- TSPAN12|NET2|TM4SF12|UNQ774/PRO1568Tetraspanin-12|Tspan-12|Tetraspan NET-2|Transmembrane 4 superfamily member 12
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95859
- O95859
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together