Anti-USP17L3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77818-113
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Ubiquitin carboxyl-terminal hydrolase 17-like protein 3 protein (19-251AA)
- Antigen Symbol:USP17L3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:A6NCW0
- Antigen Synonyms:USP17L3| USP17B| USP17F| Ubiquitin carboxyl-terminal hydrolase 17-like protein 3| EC 3.4.19.12| Deubiquitinating enzyme 17-like protein 3| Ubiquitin thioesterase 17-like protein 3| Ubiquitin-specific-processing protease 17-like protein 3
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: USP17L3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human