Anti-TNFRSF13B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TNFRSF13B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77811-221
- 77803-221
- Antigen Symbol:
- TNFRSF13B
- TNFRSF13B
- Antigen Name:
- Tumor necrosis factor receptor superfamily member 13B protein (184-293AA)
- Tumor necrosis factor receptor superfamily member 13B protein (184-293AA)
- Antigen Synonyms:
- CD 267|CD267|CD267 antigen|CVID|CVID2|FLJ39942|MGC133214|MGC39952|OTTHUMP00000065442|TNFRSF 13B|TNFRSF 14B|TNFRSF13B protein|TNFRSF14B|TR13B_HUMAN|Transmembrane activator and CAML interactor|Tumor necrosis factor receptor 13B|Tumor necrosis factor receptor superfamily member 13B
- CD 267|CD267|CD267 antigen|CVID|CVID2|FLJ39942|MGC133214|MGC39952|OTTHUMP00000065442|TNFRSF 13B|TNFRSF 14B|TNFRSF13B protein|TNFRSF14B|TR13B_HUMAN|Transmembrane activator and CAML interactor|Tumor necrosis factor receptor 13B|Tumor necrosis factor receptor superfamily member 13B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14836
- O14836
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice