Anti-PYC1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PYC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Yeast
Specifications
- Catalog No:
- 77815-168
- 77815-167
- Antigen Symbol:
- PYC1
- PYC1
- Antigen Name:
- Recombinant Baker\\\'s yeast Pyruvate carboxylase 1 protein (18-470AA)
- Recombinant Baker\\\'s yeast Pyruvate carboxylase 1 protein (18-470AA)
- Antigen Synonyms:
- PYC1|PYV|YGL062W|Pyruvate carboxylase 1|EC 6.4.1.1|Pyruvic carboxylase 1|PCB 1
- PYC1|PYV|YGL062W|Pyruvate carboxylase 1|EC 6.4.1.1|Pyruvic carboxylase 1|PCB 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Yeast
- Yeast
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P11154
- P11154
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice