Anti-RCBTB2 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: RCBTB2
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77796-911
- 77796-910
- Antigen Symbol:
- RCBTB2
- RCBTB2
- Antigen Name:
- RCC1 and BTB domain-containing protein 2 protein (1-200AA)
- RCC1 and BTB domain-containing protein 2 protein (1-200AA)
- Antigen Synonyms:
- CHC1 L|CHC1L|Chromosome condensation 1 like|RCBT2|RCBT2_HUMAN|Rcbtb2|RCC1 and BTB domain containing protein 2|RCC1 and BTB domain-containing protein 2|RCC1 like G exchanging factor|RCC1 like G exchanging factor RLG|Regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2|Regulator of chromosome condensation (RCC1) and BTB domain containing protein 2|Regulator of chromosome condensation and BTB domain containing protein 2|RLG
- CHC1 L|CHC1L|Chromosome condensation 1 like|RCBT2|RCBT2_HUMAN|Rcbtb2|RCC1 and BTB domain containing protein 2|RCC1 and BTB domain-containing protein 2|RCC1 like G exchanging factor|RCC1 like G exchanging factor RLG|Regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2|Regulator of chromosome condensation (RCC1) and BTB domain containing protein 2|Regulator of chromosome condensation and BTB domain containing protein 2|RLG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95199
- O95199
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice