Anti-IL32 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IL32
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77812-807
- 77812-806
- Antigen Symbol:
- IL32
- IL32
- Antigen Name:
- Interleukin-32 protein (31-234AA)
- Interleukin-32 protein (31-234AA)
- Antigen Synonyms:
- IL 32alpha|IL 32beta|IL 32delta|IL 32gamma|IL-32|IL32|IL32_HUMAN|Interleukin 32|Interleukin 32 small|Interleukin 32 theta|Interleukin-32|interleukin-32 eta|Natural killer cell transcript 4|Natural killer cells protein 4|NK 4|NK4|OTTHUMP00000236040|OTTHUMP00000236041|OTTHUMP00000236042|OTTHUMP00000236043|OTTHUMP00000236044|OTTHUMP00000236045|OTTHUMP00000236046|OTTHUMP00000236047|OTTHUMP00000236048|OTTHUMP00000236049|OTTHUMP00000236050|OTTHUMP00000236051|OTTHUMP00000236052|OTTHUMP00000236053|OTTHUMP00000236054|OTTHUMP00000241545|OTTHUMP00000241602|OTTHUMP00000241603|OTTHUMP00000241604|OTTHUMP00000241605|OTTHUMP00000241606|OTTHUMP00000241607|OTTHUMP00000241608|OTTHUMP00000241609|OTTHUMP00000241610|OTTHUMP00000241611|OTTHUMP00000241612|TAIF|TAIFa|TAIFb|TAIFc|TAIFd|Tumor necrosis factor alpha-inducing factor
- IL 32alpha|IL 32beta|IL 32delta|IL 32gamma|IL-32|IL32|IL32_HUMAN|Interleukin 32|Interleukin 32 small|Interleukin 32 theta|Interleukin-32|interleukin-32 eta|Natural killer cell transcript 4|Natural killer cells protein 4|NK 4|NK4|OTTHUMP00000236040|OTTHUMP00000236041|OTTHUMP00000236042|OTTHUMP00000236043|OTTHUMP00000236044|OTTHUMP00000236045|OTTHUMP00000236046|OTTHUMP00000236047|OTTHUMP00000236048|OTTHUMP00000236049|OTTHUMP00000236050|OTTHUMP00000236051|OTTHUMP00000236052|OTTHUMP00000236053|OTTHUMP00000236054|OTTHUMP00000241545|OTTHUMP00000241602|OTTHUMP00000241603|OTTHUMP00000241604|OTTHUMP00000241605|OTTHUMP00000241606|OTTHUMP00000241607|OTTHUMP00000241608|OTTHUMP00000241609|OTTHUMP00000241610|OTTHUMP00000241611|OTTHUMP00000241612|TAIF|TAIFa|TAIFb|TAIFc|TAIFd|Tumor necrosis factor alpha-inducing factor
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24001
- P24001
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice