Anti-SMC1B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: SMC1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77833-444
- 77833-443
- Antigen Symbol:
- SMC1B
- SMC1B
- Antigen Name:
- Structural maintenance of chromosomes protein 1B protein (954-1060AA)
- Structural maintenance of chromosomes protein 1B protein (954-1060AA)
- Antigen Synonyms:
- bK268H5|SMC protein 1B|SMC-1-beta|SMC-1B|SMC1 beta|SMC1B|SMC1B_HUMAN|SMC1beta|SMC1beta protein|Smc1l2|Structural maintenance of chromosomes 1-like 2|Structural maintenance of chromosomes 1B|Structural maintenance of chromosomes protein 1B
- bK268H5|SMC protein 1B|SMC-1-beta|SMC-1B|SMC1 beta|SMC1B|SMC1B_HUMAN|SMC1beta|SMC1beta protein|Smc1l2|Structural maintenance of chromosomes 1-like 2|Structural maintenance of chromosomes 1B|Structural maintenance of chromosomes protein 1B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8NDV3
- Q8NDV3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together