Anti-SIPA1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SIPA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-606
- 77799-607
- Antigen Symbol:
- SIPA1
- SIPA1
- Antigen Name:
- Signal-induced proliferation-associated protein 1 protein (759-927AA)
- Signal-induced proliferation-associated protein 1 protein (759-927AA)
- Antigen Synonyms:
- GTPase activating protein Spa 1|GTPase-activating protein Spa-1|MGC102688|MGC17037|p130 SPA-1|p130 SPA1|Signal induced proliferation associated 1|Signal induced proliferation associated gene 1|Signal induced proliferation associated protein 1|Signal-induced proliferation-associated protein 1|SIPA 1|Sipa-1|Sipa1|SIPA1_HUMAN|Spa1
- GTPase activating protein Spa 1|GTPase-activating protein Spa-1|MGC102688|MGC17037|p130 SPA-1|p130 SPA1|Signal induced proliferation associated 1|Signal induced proliferation associated gene 1|Signal induced proliferation associated protein 1|Signal-induced proliferation-associated protein 1|SIPA 1|Sipa-1|Sipa1|SIPA1_HUMAN|Spa1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FS4
- Q96FS4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice