Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MSRA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-MSRA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MSRA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77841-627
  • 77841-628
  • Antigen Symbol:
  • MSRA
  • MSRA
  • Antigen Name:
  • Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
  • Mitochondrial peptide methionine sulfoxide reductase protein (24-235AA)
  • Antigen Synonyms:
  • Cytosolic methionine S sulfoxide reductase| Methionine sulphoxide reductase A| Mitochondrial peptide methionine sulfoxide reductase| MSR A| msrA| _HUMAN| peptide met (O) reductase| Peptide Met(O) reductase| Peptide methionine (S) S oxide reductase| Peptide-methionine (S)-S-oxide reductase| PMSR| Protein methionine S oxide reductase| Protein-methionine-S-oxide reductase
  • Cytosolic methionine S sulfoxide reductase| Methionine sulphoxide reductase A| Mitochondrial peptide methionine sulfoxide reductase| MSR A| msrA| _HUMAN| peptide met (O) reductase| Peptide Met(O) reductase| Peptide methionine (S) S oxide reductase| Peptide-methionine (S)-S-oxide reductase| PMSR| Protein methionine S oxide reductase| Protein-methionine-S-oxide reductase
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UJ68
  • Q9UJ68
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications