Anti-EPPIN Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: EPPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-814
- 77819-813
- Antigen Symbol:
- EPPIN
- EPPIN
- Antigen Name:
- Synthetic peptide of EPPIN
- Synthetic peptide of EPPIN
- Antigen Synonyms:
- Cancer/testis antigen 71|CT 71|CT71|dJ461P17.2|Epididymal protease inhibitor|EPPI_HUMAN|EPPIN 1|EPPIN 2|EPPIN 3|EPPIN|EPPIN1|EPPIN2|EPPIN3|Protease inhibitor WAP7|Serine peptidase inhibitor like with Kunitz and WAP domains 1 (eppin)|Serine peptidase inhibitor like with Kunitz and WAP domains 1|Serine protease inhibitor-like with Kunitz and WAP domains 1|SPINLW 1|WAP 7|WAP four disulfide core domain 7|WAP four disulfide core domain protein 7|WAP four-disulfide core domain protein 7|WAP7|WFDC 7|WFDC7
- Cancer/testis antigen 71|CT 71|CT71|dJ461P17.2|Epididymal protease inhibitor|EPPI_HUMAN|EPPIN 1|EPPIN 2|EPPIN 3|EPPIN|EPPIN1|EPPIN2|EPPIN3|Protease inhibitor WAP7|Serine peptidase inhibitor like with Kunitz and WAP domains 1 (eppin)|Serine peptidase inhibitor like with Kunitz and WAP domains 1|Serine protease inhibitor-like with Kunitz and WAP domains 1|SPINLW 1|WAP 7|WAP four disulfide core domain 7|WAP four disulfide core domain protein 7|WAP four-disulfide core domain protein 7|WAP7|WFDC 7|WFDC7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95925
- O95925
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice