Anti-GCSAML Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: GCSAML
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77827-055
- 77827-056
- Antigen Symbol:
- GCSAML
- GCSAML
- Antigen Name:
- Germinal center-associated signaling and motility-like protein (1-135AA)
- Germinal center-associated signaling and motility-like protein (1-135AA)
- Antigen Synonyms:
- GCSAML|C1orf150|Germinal center-associated signaling and motility-like protein
- GCSAML|C1orf150|Germinal center-associated signaling and motility-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5JQS6
- Q5JQS6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice