Anti-PRKAB2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77819-439
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:5\\\'-AMP-activated protein kinase subunit beta-2 protein (1-272AA)
- Antigen Symbol:PRKAB2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O43741
- Antigen Synonyms:5' AMP activated protein kinase beta 2 subunit|5' AMP activated protein kinase subunit beta 2|5''-AMP-activated protein kinase subunit beta-2|AAKB2_HUMAN|AMP activated protein kinase beta 2 non catalytic subunit|AMPK beta 2|AMPK beta 2 chain|AMPK subunit beta 2|AMPK subunit beta-2|MGC61468|PRKAB 2|Prkab2|Protein kinase AMP activated beta 2 non catalytic subunit
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: PRKAB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat