Anti-PPP2R2C Rabbit Polyclonal Antibody
Catalog # 77795-124
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform
- Antigen Symbol:PPP2R2C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9Y2T4
- Antigen Synonyms:2ABG_HUMAN|B55 gamma|Gamma isoform of regulatory subunit B55 protein phosphatase 2|IMYPNO 1|IMYPNO|IMYPNO1|MGC33570|OTTHUMP00000115505|Phosphoprotein phosphatase 2A BR gamma regulatory chain|PP2A subunit B B gamma isoform|PP2A subunit B B55 gamma isoform|PP2A subunit B isoform B55-gamma|PP2A subunit B isoform gamma|PP2A subunit B isoform PR55-gamma|PP2A subunit B isoform R2-gamma|PP2A subunit B PR55 gamma isoform|PP2A subunit B R2 gamma isoform|PPP2R2C|PPP2R2C protein|PR 52|PR 55G|PR52|PR55G|Protein phosphatase 2 (formerly 2A) regulatory subunit B gamma isoform|Protein phosphatase 2 regulatory subunit B gamma|Protein phosphatase 2 regulatory subunit B gamma isoform|Protein phosphatase 2A1 B gamma subunit|Serine/threonine protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform|Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B gamma isoform
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: PPP2R2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat